ADVH49B83162 AChemBlock

methyl (R)-3-amino-2-phenylpropanoate hydrochloride

  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/methyl-r-3-amino-2-phenylpropanoate-hydrochloride-310559.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87250
Availability details for
Quantity
  • Availability
  • Add to Cart