ADVH49B83164 AChemBlock

6-chloro-1H-indole-4-carbonitrile

  • Cas Number : 1097629-20-3
  • Molecular Weight : 176.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-1h-indole-4-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87299
Availability details for
Quantity
  • Availability
  • Add to Cart