ADVH49B83167 AChemBlock

methyl 6-methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate

  • Cas Number : 872355-54-9
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-methyl-1h-pyrrolo-2-3-b-pyridine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87302
Availability details for
Quantity
  • Availability
  • Add to Cart