ADVH49B83169 AChemBlock

tert-butyl (E)-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)allyl)carbamate

  • Cas Number : 468060-28-8
  • Molecular Weight : 283.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-e-3-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-allyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72330
Availability details for
Quantity
  • Availability
  • Add to Cart