ADVH49B8316A AChemBlock

MLN120B

  • Cas Number : 1256790-93-8
  • Molecular Weight : 366.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/mln120b.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87306
Availability details for
Quantity
  • Availability
  • Add to Cart