ADVH49B8316E AChemBlock

methyl 4-(1-aminocyclopropyl)benzoate hydrochloride

  • Cas Number : 1014645-87-4
  • Molecular Weight : 227.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-1-aminocyclopropyl-benzoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87331
Availability details for
Quantity
  • Availability
  • Add to Cart