ADVH49B8316F AChemBlock

(R)-pentan-2-amine hydrochloride

  • Cas Number : 101689-05-8
  • Molecular Weight : 123.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-pentan-2-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87332
Availability details for
Quantity
  • Availability
  • Add to Cart