ADVH49B83170 AChemBlock

(S)-6-fluorochroman-4-amine

  • Cas Number : 1018978-85-2
  • Molecular Weight : 167.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-6-fluorochroman-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87333
Availability details for
Quantity
  • Availability
  • Add to Cart