ADVH49B83174 AChemBlock

7-methylisochromane

  • Cas Number : 103261-92-3
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-methylisochromane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87337
Availability details for
Quantity
  • Availability
  • Add to Cart