ADVH49B83178 AChemBlock

(R)-2-amino-2-(2-methoxyphenyl)acetic acid hydrochloride

  • Cas Number : 103889-79-8
  • Molecular Weight : 217.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-amino-2-2-methoxyphenyl-acetic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87341
Availability details for
Quantity
  • Availability
  • Add to Cart