ADVH49B8317A AChemBlock

2-((tert-butoxycarbonyl)amino)-3-(naphthalen-1-yl)propanoic acid

  • Cas Number : 104882-22-6
  • Molecular Weight : 315.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-tert-butoxycarbonyl-amino-3-naphthalen-1-yl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87343
Availability details for
Quantity
  • Availability
  • Add to Cart