ADVH49B8317C AChemBlock

1-(6-methoxypyridin-2-yl)ethan-1-amine

  • Cas Number : 1060807-26-2
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-6-methoxypyridin-2-yl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87345
Availability details for
Quantity
  • Availability
  • Add to Cart