ADVH49B83181 AChemBlock

(R)-1-(3-fluoro-4-(trifluoromethyl)phenyl)ethan-1-amine

  • Cas Number : 1079656-75-9
  • Molecular Weight : 207.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-3-fluoro-4-trifluoromethyl-phenyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87350
Availability details for
Quantity
  • Availability
  • Add to Cart