ADVH49B83182 AChemBlock

6-methyl-3,4-dihydroisoquinolin-1(2H)-one

  • Cas Number : 1082041-78-8
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-methyl-3-4-dihydroisoquinolin-1-2h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87351
Availability details for
Quantity
  • Availability
  • Add to Cart