ADVH49B83186 AChemBlock

1-(2,6-dimethoxyphenyl)ethan-1-amine hydrochloride

  • Cas Number : 1087707-43-4
  • Molecular Weight : 217.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-6-dimethoxyphenyl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87355
Availability details for
Quantity
  • Availability
  • Add to Cart