ADVH49B83187 AChemBlock

3-(4-bromophenyl)-1-methylpyrrolidine

  • Cas Number : 1088410-99-4
  • Molecular Weight : 240.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-4-bromophenyl-1-methylpyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87356
Availability details for
Quantity
  • Availability
  • Add to Cart