ADVH49B83192 AChemBlock

7-bromo-8-fluorochroman-4-one

  • Cas Number : 1092350-77-0
  • Molecular Weight : 245.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-8-fluorochroman-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87367
Availability details for
Quantity
  • Availability
  • Add to Cart