ADVH49B83194 AChemBlock

1-(4-fluorophenyl)propan-1-amine hydrochloride

  • Cas Number : 1092797-76-6
  • Molecular Weight : 189.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-fluorophenyl-propan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87369
Availability details for
Quantity
  • Availability
  • Add to Cart