ADVH49B8319A AChemBlock

6-(trifluoromethyl)chroman-4-one

  • Cas Number : 1122410-37-0
  • Molecular Weight : 216.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-trifluoromethyl-chroman-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87375
Availability details for
Quantity
  • Availability
  • Add to Cart