ADVH49B8319C AChemBlock

2-(aminomethyl)benzonitrile hydrochloride

  • Cas Number : 1134529-25-1
  • Molecular Weight : 168.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-aminomethyl-benzonitrile-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87377
Availability details for
Quantity
  • Availability
  • Add to Cart