ADVH49B831A0 AChemBlock

2-amino-2-(3,5-dichlorophenyl)acetic acid hydrochloride

  • Cas Number : 1137014-87-9
  • Molecular Weight : 256.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-2-3-5-dichlorophenyl-acetic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87381
Availability details for
Quantity
  • Availability
  • Add to Cart