ADVH49B831A1 AChemBlock

(R)-1-(2-bromophenyl)ethan-1-amine

  • Cas Number : 113974-24-6
  • Molecular Weight : 200.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-2-bromophenyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87382
Availability details for
Quantity
  • Availability
  • Add to Cart