ADVH49B831A4 AChemBlock

1-(3-bromo-5-fluorophenyl)-2-methylpropan-1-one

  • Cas Number : 1147871-74-6
  • Molecular Weight : 245.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-bromo-5-fluorophenyl-2-methylpropan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87385
Availability details for
Quantity
  • Availability
  • Add to Cart