ADVH49B831A8 AChemBlock

(2-(aminomethyl)pyridin-3-yl)methanol dihydrochloride

  • Cas Number : 1159812-22-2
  • Molecular Weight : 211.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-aminomethyl-pyridin-3-yl-methanol-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87389
Availability details for
Quantity
  • Availability
  • Add to Cart