ADVH49B831AA AChemBlock

methyl 8-oxo-5,6,7,8-tetrahydronaphthalene-2-carboxylate

  • Cas Number : 116047-26-8
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-8-oxo-5-6-7-8-tetrahydronaphthalene-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87391
Availability details for
Quantity
  • Availability
  • Add to Cart