ADVH49B831AC AChemBlock

1-(2-fluorophenyl)-2-methylpropan-1-one

  • Cas Number : 116707-09-6
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-fluorophenyl-2-methylpropan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87393
Availability details for
Quantity
  • Availability
  • Add to Cart