ADVH49B831AD AChemBlock

(R)-1-(3-fluorophenyl)propan-1-amine hydrochloride

  • Cas Number : 1168139-41-0
  • Molecular Weight : 189.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-3-fluorophenyl-propan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87394
Availability details for
Quantity
  • Availability
  • Add to Cart