ADVH49B831B9 AChemBlock

2-(2,4-difluorophenyl)pyrrolidine hydrochloride

  • Cas Number : 1189996-39-1
  • Molecular Weight : 219.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-4-difluorophenyl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87406
Availability details for
Quantity
  • Availability
  • Add to Cart