ADVH49B831BC AChemBlock

7-bromo-2,3-dihydro-1H-inden-1-ol

  • Cas Number : 1196049-18-9
  • Molecular Weight : 213.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-2-3-dihydro-1h-inden-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87409
Availability details for
Quantity
  • Availability
  • Add to Cart