ADVH49B831BE AChemBlock

2-(4-(trifluoromethyl)phenyl)pyrrolidine hydrochloride

  • Cas Number : 1197232-77-1
  • Molecular Weight : 251.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-trifluoromethyl-phenyl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87411
Availability details for
Quantity
  • Availability
  • Add to Cart