ADVH49B831C2 AChemBlock

5-bromochroman-4-one

  • Cas Number : 1199782-67-6
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromochroman-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87415
Availability details for
Quantity
  • Availability
  • Add to Cart