ADVH49B831C3 AChemBlock

7-chloro-5-hydroxy-2,3-dihydro-1H-inden-1-one

  • Cas Number : 1199782-69-8
  • Molecular Weight : 182.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-chloro-5-hydroxy-2-3-dihydro-1h-inden-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87416
Availability details for
Quantity
  • Availability
  • Add to Cart