ADVH49B831CA AChemBlock

methyl 2-(pyrrolidin-2-yl)benzoate hydrochloride

  • Cas Number : 1203681-57-5
  • Molecular Weight : 241.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-pyrrolidin-2-yl-benzoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87423
Availability details for
Quantity
  • Availability
  • Add to Cart