ADVH49B831CC AChemBlock

methyl 4-bromo-5-(difluoromethyl)thiophene-2-carboxylate

  • Cas Number : 1207557-47-8
  • Molecular Weight : 271.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-bromo-5-difluoromethyl-thiophene-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87425
Availability details for
Quantity
  • Availability
  • Add to Cart