ADVH49B831CD AChemBlock

2,2-difluorobenzo[d][1,3]dioxol-5-ol

  • Cas Number : 1211539-82-0
  • Molecular Weight : 174.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-difluorobenzo-d-1-3-dioxol-5-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87426
Availability details for
Quantity
  • Availability
  • Add to Cart