ADVH49B831D3 AChemBlock

(S)-2-(3-chloro-5-fluorophenyl)piperidine

  • Cas Number : 1212955-17-3
  • Molecular Weight : 213.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-3-chloro-5-fluorophenyl-piperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87432
Availability details for
Quantity
  • Availability
  • Add to Cart