ADVH49B831D4 AChemBlock

(R)-1-(5-fluoro-2-methoxypyridin-3-yl)ethan-1-amine

  • Cas Number : 1212996-56-9
  • Molecular Weight : 170.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-5-fluoro-2-methoxypyridin-3-yl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87433
Availability details for
Quantity
  • Availability
  • Add to Cart