ADVH49B831D6 AChemBlock

(S)-1-(4-(cyclopentyloxy)phenyl)ethan-1-amine

  • Cas Number : 1213104-18-7
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-4-cyclopentyloxy-phenyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87435
Availability details for
Quantity
  • Availability
  • Add to Cart