ADVH49B831D7 AChemBlock

(S)-2-amino-2-(o-tolyl)ethan-1-ol hydrochloride

  • Cas Number : 1213107-41-5
  • Molecular Weight : 187.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-2-o-tolyl-ethan-1-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87436
Availability details for
Quantity
  • Availability
  • Add to Cart