ADVH49B831DA AChemBlock

(R)-1-(2,2-difluorobenzo[d][1,3]dioxol-4-yl)ethan-1-amine

  • Cas Number : 1213203-92-9
  • Molecular Weight : 201.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-2-2-difluorobenzo-d-1-3-dioxol-4-yl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87439
Availability details for
Quantity
  • Availability
  • Add to Cart