ADVH49B831DB AChemBlock

(S)-1-(4-fluoro-3-methylphenyl)ethan-1-amine

  • Cas Number : 1213299-63-8
  • Molecular Weight : 153.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-4-fluoro-3-methylphenyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87440
Availability details for
Quantity
  • Availability
  • Add to Cart