ADVH49B831DC AChemBlock

(S)-2-amino-2-(3,5-dimethylphenyl)ethan-1-ol

  • Cas Number : 1213317-54-4
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-2-3-5-dimethylphenyl-ethan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87441
Availability details for
Quantity
  • Availability
  • Add to Cart