ADVH49B831E3 AChemBlock

(S)-1-(6-bromopyridin-3-yl)ethan-1-amine

  • Cas Number : 1213594-37-6
  • Molecular Weight : 201.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-6-bromopyridin-3-yl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87448
Availability details for
Quantity
  • Availability
  • Add to Cart