ADVH49B831E4 AChemBlock

(S)-8-methylchroman-4-amine

  • Cas Number : 1213596-97-4
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-8-methylchroman-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87449
Availability details for
Quantity
  • Availability
  • Add to Cart