ADVH49B831E7 AChemBlock

6-bromo-2,3-dihydrobenzofuran-3-amine hydrochloride

  • Cas Number : 1215339-86-8
  • Molecular Weight : 250.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-3-dihydrobenzofuran-3-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87452
Availability details for
Quantity
  • Availability
  • Add to Cart