ADVH49B831E9 AChemBlock

methyl (R)-3-amino-3-(4-chlorophenyl)propanoate hydrochloride

  • Cas Number : 1217530-86-3
  • Molecular Weight : 250.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-r-3-amino-3-4-chlorophenyl-propanoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87454
Availability details for
Quantity
  • Availability
  • Add to Cart