ADVH49B831EC AChemBlock

(S)-2-(4-chlorophenyl)pyrrolidine

  • Cas Number : 1217651-75-6
  • Molecular Weight : 181.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-4-chlorophenyl-pyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87457
Availability details for
Quantity
  • Availability
  • Add to Cart