ADVH49B831F1 AChemBlock

(R)-2-(2,5-dichlorophenyl)pyrrolidine

  • Cas Number : 1228569-01-4
  • Molecular Weight : 216.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-2-5-dichlorophenyl-pyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87462
Availability details for
Quantity
  • Availability
  • Add to Cart