ADVH49B831F8 AChemBlock

methyl (S)-3-aminopentanoate hydrochloride

  • Cas Number : 1236525-13-5
  • Molecular Weight : 167.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-s-3-aminopentanoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87469
Availability details for
Quantity
  • Availability
  • Add to Cart