ADVH49B831FD AChemBlock

(R)-3-(1-amino-2-hydroxyethyl)benzonitrile hydrochloride

  • Cas Number : 1245623-77-1
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-1-amino-2-hydroxyethyl-benzonitrile-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87474
Availability details for
Quantity
  • Availability
  • Add to Cart